
Potassium permanganate 6% saturated solution in water for the determination of chloride in cheese acc. Erbacher
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7722-64-7
:
Formula:KMnO₄
MDL Number:MFCD00011364
:158.03 g/mol
Storage Temperature:Ambient
EINECS:231-760-3
UN:3082
ADR:9,III
:GHS09