Organic reference standard, 2-Isobutyl-3-methoxypyrazine 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA14394700ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:24683-00-9
- Description:2-Isobutyl-3-methoxypyrazine 100 µg/ml in methanol
- Formula:C₁₂H₂₀O₂
- Description:2-Isobutyl-3-methoxypyrazine