Order Entry
Germany
ContactUsLinkComponent
1-Octadecanethiol for synthesis
1-Octadecanethiol for synthesis
Catalog # KRAF19756.0801
Supplier:  Bernd Kraft
CAS Number:  
1-Octadecanethiol for synthesis
Catalog # KRAF19756.0801
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :286.57 g/mol
Boiling Pt:204 °C (1013 hPa)
Melting Pt:28…31 °C
Density:0.8475 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:220-744-1
UN:3335
ADR:9,
CAS Number:2885-00-9
Formula:C₁₈H₃₈S
MDL Number:MFCD00004886