Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Distillation Column Packing
Raschig rings
Raschig rings
Raschig rings
Supplier:
GLASWARENFABRIK KARL HECHT
Certificates
Expand image
Raschig rings
Supplier:
GLASWARENFABRIK KARL HECHT
About this item
Soda lime glass.
For packing columns
Good chemical resistance
Good thermal resistance