Super syringes
Supplier: HAMILTON BONADUZ
Certificates
About this item
Super syringes with PTFE Luer lock (TLL) connector or tracheal adapter.
- Clear acrylic syringe barrels
- For use with gases and liquids
Ideal for use with air sampling, preparing gas standards, calibrating reservoirs, and pneumographs.