Clamps, three pronged
Catalog # WITG8.430.000
Supplier: witeg Labortechnik
Specifications
- Spanning width:0 - 120 mm
- Pk:1
Specifications
About this item
These universal three-pronged clamps are made of laquered aluminium, with nickel plated brass screws.
- Jaws are covered with a layer of cork to protect clamped items