Order Entry
Germany
ContactUsLinkComponent
Acetic acid 50%
Acetic acid 50%
Catalog # 86003.980
Supplier:  VWR Collection
CAS Number:  
undefined
Acetic acid 50%
Catalog # 86003.980
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pk:
    950 L
  • Seq:
    5783

Specifications

About this item

 :
 :60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2790
ADR:8,II
 :GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152