Cans, waste disposal
Supplier: KGW
Certificates
About this item
Insert made of borosilicate glass 3.3. Ideal for storage and transport of aggressive waste in laboratories.
- Good tipping stability
- Glass is resistant to all kind of chemicals
- Very good temperature resistance
Certifications: In accordance with DIN 12111, DIN 12116, DIN 52322.