Benzo[k]fluoranthene
Catalog # PROOBCR-048R
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
EINECS:205-916-6
CAS Number:207-08-9
Formula:C₂₀H₁₂
MDL Number:MFCD00046287