2-Chloro-1,3,2-benzodioxaborole (β-bromocatecholborane), Sigma-Aldrich®
Catalog # SIAL362700-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
CAS Number:55718-76-8
Formula:C₆H₄BClO₂
MDL Number:MFCD00043395