Order Entry
Germany
ContactUsLinkComponent
 
Pesticide reference standard, Trichloronat (O-Ethyl O-2,4,5-trichlorophenyl ethylphosphonothioate) 100 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERXA17750000CY
 :  
Pesticide reference standard, Trichloronat (O-Ethyl O-2,4,5-trichlorophenyl ethylphosphonothioate) 100 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERXA17750000CY
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    327-98-0
  • Description:
    Trichloronat (O-Ethyl O-2,4,5-trichlorophenyl ethylphosphonothioate) 100 µg/ml in cyclohexane
  • Formula:
    C₁₀H₁₂Cl₃O₂PS
  • Description:
    Trichloronat (O-Ethyl O-2,4,5-trichlorophenyl ethylphosphonothioate)