Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Dimethylamino)-4'-fluorobenzhydrol
4-(Dimethylamino)-4'-fluorobenzhydrol
4-(Dimethylamino)-4'-fluorobenzhydrol
4-(Dimethylamino)-4'-fluorobenzhydrol
Catalog # FLUO022515-1G
Supplier:
FLUOROCHEM
CAS Number:
Expand image
4-(Dimethylamino)-4'-fluorobenzhydrol
Catalog # FLUO022515-1G
Supplier:
FLUOROCHEM
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
Specifications
About this item
Product Details
:
CAS Number:
39768-80-4
Formula:
C₁₅H₁₆FNO
Product Details