Order Entry
Germany
ContactUsLinkComponent
2,2-Diphenylthioacetamide
2,2-Diphenylthioacetamide
Catalog # FLUO021351-1G
Supplier:  FLUOROCHEM
CAS Number:  
2,2-Diphenylthioacetamide
Catalog # FLUO021351-1G
Supplier:  FLUOROCHEM
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
CAS Number:17518-50-2
Formula:C₁₄H₁₃NS
MDL Number:MFCD01670747