2-Chloro-2,2-dfluoroacetyl chloride
Catalog # FLUO004393-5G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
EINECS:206-551-5
CAS Number:354-24-5
Formula:C₂Cl₂F₂O
MDL Number:MFCD00039317