Order Entry
Germany
ContactUsLinkComponent
 
2-Mercaptobenzoxazole 99%
  ACRO125460250
 :  
undefined
2-Mercaptobenzoxazole 99%
  ACRO125460250
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    192 to 195 °C
  • Titration Argentometric:
    ≥98.5 %
  • HPLC:
    ≥98.5 %

 

 

 :
 :151.19 g/mol
 :192…195 °C
 :Ambient
 :219-191-9
 :2382-96-9
 :C₇H₅NOS
 :MFCD00005769