
Sulphuric acid-[D₂] 98% (99.5% D) (w/w) in D₂O for NMR spectroscopy, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13813-19-9
:
Formula:D₂SO₄
MDL Number:MFCD00003542
:100.09 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:237-464-0
UN:1830
:GHS05
Storage Temperature:Ambient