
Sulphuric acid-[D₂] 96% (99.5% D) (w/w) in D₂O for NMR spectroscopy
Catalog # 87857.18
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
Specifications
About this item
:
:100.09 g/mol
Boiling Pt:290 °C
Density:1.86
Storage Temperature:Ambient
EINECS:237-464-0
UN:1830
ADR:8,II
:GHS05
CAS Number:13813-19-9
Formula:D₂SO₄
MDL Number:MFCD00003542