Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-2,6-dimethoxypyrimidine
4-Chloro-2,6-dimethoxypyrimidine ≥98%
4-Chloro-2,6-dimethoxypyrimidine ≥98%
:
Thermo Fisher Scientific
4-Chloro-2,6-dimethoxypyrimidine ≥98%
:
Thermo Fisher Scientific
:
6320-15-6
:
:
C₆H₇ClN₂O₂
:
MFCD00006068
:
174.59 g/mol
:
71…73 °C
:
228-669-6
:
GHS07
:
Ambient
msds_ALFAB21696
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years