Azidotrimethylsilane for synthesis, Sigma-Aldrich®
8.18026.0025
: Merck
:
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
- Solubility:(20 °C) (decomposition)
- Molar Mass:115.21 g/mol
- Boiling Point:92 - 95 °C
- Flash Point:6 °C
- Refractive Index:1.4142 (20 °C, 589 nm)
- Density:0.88 g/cm3 (20 °C)
- Ignition Point:300 °C
:
:115.21 g/mol
:3 (6.1),II
:M-Clarity™ quality level = MQ200
:225-078-5
:1992
:GHS02
:GHS06
:Ambient
:4648-54-8
:C₃H₉N₃Si
:MFCD00001986