Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethosuximide
Ethosuximide, Sigma-Aldrich®
Ethosuximide, Sigma-Aldrich®
Ethosuximide, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Ethosuximide, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
77-67-8
:
Formula:
C₇H₁₁NO₂
MDL Number:
MFCD00072123
:
141.17 g/mol
:
M-Clarity™ quality level = MQ100
EINECS:
201-048-7