Order Entry
Germany
ContactUsLinkComponent
Supelco® Organic Reference Standard, Digoxin
Supelco® Organic Reference Standard, Digoxin
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, Digoxin
Supplier:  Merck

Specifications

  • CAS No.:
    20830-75-5
  • Storage conditions:
    RT
  • Hazard class description:
    H300 - H331 - H373
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00003674
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    12β-Hydroxydigitoxin
  • Molecular weight:
    780,94

Specifications

Frequently Bought Together