Digoxin
Glass bottle for solids, 1 mg
Catalog # FITZ10-D05-M94146
Supplier: Fitzgerald Industries
Jump to:
Product Details & Documents
- CAS Number:20830-75-5
- Formula:C₄₁H₆₄O₁₄
- MDL Number:MFCD00003674
- Molecular Weight:780.95 g/mol
- EINECS:244-068-1
- UN:2811
- ADR:6.1,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



