Order Entry
Germany
Orders LinkContactUsLinkComponent
Digoxin

Digoxin

Glass bottle for solids, 1 mg
Catalog # FITZ10-D05-M91286
Digoxin
Catalog # FITZ10-D05-M91286
CAS Number:  20830-75-5
Digoxin

Glass bottle for solids, 1 mg

CAS Number:  20830-75-5
Price
Inquire For Price
Log in for availability
1 * 1 mg
Product Details & Documents
  • CAS Number:20830-75-5
  • Formula:C₄₁H₆₄O₁₄
  • MDL Number:MFCD00003674
  • Molecular Weight:780.95 g/mol
  • EINECS:244-068-1
  • UN:2811
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 mg

Specifications