Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,5-Dibromo-2,2,3,3,4,4-hexafluoropentane
1,5-Dibromo-2,2,3,3,4,4-hexafluoropentane
1,5-Dibromo-2,2,3,3,4,4-hexafluoropentane
1,5-Dibromo-2,2,3,3,4,4-hexafluoropentane
Supplier:
FLUOROCHEM
Certificates
Expand image
1,5-Dibromo-2,2,3,3,4,4-hexafluoropentane
Supplier:
FLUOROCHEM
About this item
CAS Number:
33619-78-2
:
Formula:
C₅H₄Br₂F₆
MDL Number:
MFCD03095411