Skip to Header
Skip to Main Content
Skip to Footer
Erweiterte Suche
Erweiterte Suche
Sicherheitsdatenblatt (SDS)
Zertifikate
Herstellerdaten
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane
1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane ≥97%
1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane ≥97%
Irritant
1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
371-83-5
:
Formula:
C₅H₄Br₂F₆
MDL Number:
MFCD00236657
:
337.89 g/mol
Density:
2.003
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAB21405
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation