2-Aminoethyl dihydrogen phosphate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1071-23-4
:
Formula:C₂H₈NO₄P
MDL Number:MFCD00008178
:141.0630 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:213-988-5
UN:3261
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:241 - 243 °C
- Molar Mass:141.06 g/mol
- Synonyms:Colamine phosphoric acid, Ethanolamine dihydrogen phosphate, 2-Hydroxyethylammonium dihydrogen phosphate, O-Phosphoethanolamine
- Hill Formula:C2H8NO4P
- Chemical Formula:H2NCH2CH2OPO(OH)2