Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dichlorobenzyl mercaptan
2,4-Dichlorobenzyl mercaptan ≥98%
2,4-Dichlorobenzyl mercaptan ≥98%
:
Thermo Fisher Scientific
2,4-Dichlorobenzyl mercaptan ≥98%
:
Thermo Fisher Scientific
:
59293-67-3
:
:
C₇H₆Cl₂S
:
MFCD00039650
:
193.10 g/mol
:
150…151 °C (12 mmHg)
:
1.36
:
>100 °C (212 °F)
:
Ambient
:
261-687-2
:
2810
:
6.1,III
:
GHS06
:
GHS07
msds_ALFAB24999
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years