3,4-Dichloro-3'-methylbenzhydrol
Catalog # FLUO022438-1G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
CAS Number:842140-73-2
Formula:C₁₄H₁₂Cl₂O
MDL Number:MFCD06201268