Order Entry
Germany
ContactUsLinkComponent
Potassium cyanide 100 g/l in aqueous solution
Potassium cyanide 100 g/l in aqueous solution
Catalog # RICC6040-16
Supplier:  Ricca Chemical
CAS Number:  
Potassium cyanide 100 g/l in aqueous solution
Catalog # RICC6040-16
Supplier:  Ricca Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :65.12 g/mol
Storage Temperature:Ambient
EINECS:205-792-3
UN:3413
ADR:6.1,II
CAS Number:151-50-8
Formula:KCN
MDL Number:MFCD00011397