Order Entry
Germany
Orders LinkContactUsLinkComponent
Methanol

Methanol

5 L
Catalog # LBCL5L561200
Flammable
Toxic
Health
Marketsource
Supplier:   LABC LABORTECHNIK
Methanol
Catalog # LBCL5L561200
Supplier:  LABC LABORTECHNIK
Marketsource
CAS Number:  67-56-1
Methanol
Methanol

5 L

CAS Number:  67-56-1
Price
130,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
  • CAS Number:67-56-1
  • Formula:CH₃OH
  • MDL Number:MFCD00004595
  • Molecular Weight:32.04 g/mol
  • Merck Index:13,05984
  • Boiling Pt:64.6 °C (1013 hPa)
  • Melting Pt:–98 °C
  • Density:0.7918 g/cm³ (20 °C)
  • Flash Pt:11 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-659-6
  • UN:1230
  • ADR:3,II
  • REACH:01-2119433307-44
Specifications

Specifications

  • Pk:
    5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.