Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Safranine O

Glass bottle for solids, 0,1 g
Catalog # EHERC10424960
Supplier:   EHRENSTORFER
Safranine O
Catalog # EHERC10424960
Supplier:  EHRENSTORFER
CAS Number:  477-73-6
Safranine O

Glass bottle for solids, 0,1 g

CAS Number:  477-73-6
Price
Inquire For Price
Log in for availability
1 * 0,1 g
Product Details & Documents
Synonyms:  Gossypimine, Safranin O, Safranin Y, Safranin A, 3,7-Diamino-2,8-dimethyl-5-phenylphenazinium chloride, 3,7-Diamino-2,8-dimethyl-5-phenylphenazin-5-ium chloride, Safranin T, CI 50240, Basic Red 2, Safranin, Cotton Red
  • CAS Number:477-73-6
  • Formula:C₂₀H₁₉ClN₄
  • MDL Number:MFCD00011759
  • Molecular Weight:350.85 g/mol
  • Density:1.447 g/cm³ (22 °C)
  • Storage Temperature:Ambient
  • EINECS:207-518-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g

Specifications