Order Entry
Germany
Orders LinkContactUsLinkComponent
3-(2-Fluorophenyl)propionic acid 96%

3-(2-Fluorophenyl)propionic acid 96%

3-(2-Fluorophenyl)propionic acid 96%
Certificates
3-(2-Fluorophenyl)propionic acid 96%
3-(2-Fluorophenyl)propionic acid 96%

3 Options Available Below

Product Details & Documents

Purity: 96% CAS No.: 1643-26-1 Molecular Formula: C9H9FO2 Formula Weight: 168.17 Melting Point: 75-79° MDL No.: MFCD01310820

  • CAS Number:1643-26-1
  • Formula:C₉H₉FO₂
  • MDL Number:MFCD01310820
  • Molecular Weight:168.17 g/mol
  • Melting Pt:75…79 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.