Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fullerene C60
Fullerene C60 99%, powder
Fullerene C60 99%, powder
:
Thermo Fisher Scientific
Fullerene C60 99%, powder
:
Thermo Fisher Scientific
:
99685-96-8
:
:
C₆₀
:
MFCD00151408
:
13,01447
:
500…600 °C (subl.)
:
> 280 °C
:
1.7
:
1325
:
4.1,III
:
720.66 g/mol
:
GHS02
:
GHS07
:
Ambient
msds_ALFA046311
Thermo Scientific Acroseal 25 Years