Protective hand creams, Baktolan® Protect+ Pure
Supplier: BODE CHEMIE
Certificates
About this item
Baktolan® Protect+ Pure is used where highly stressed or even cracked skin needs to be regenerated and strengthened. It protects the skin when performing tasks with water-soluble substances.
- Replenishing w/o/w emulsion for the protection against aqueous solutions and for dry, highly stressed skin
- Strengthens the skin barrier
- Prevents skin irritations that might be caused by using gloves
- Free from preservatives, fragrance, silicone, and colourants
Particularly suitable for daily application in hospitals and health care, nursing, chemical, pharamceutical, and food processing areas.