Order Entry
Germany
Orders LinkContactUsLinkComponent
Bottles, water sampling

Bottles, water sampling

250 ml, Undosed, bulk
Catalog # LPIT292258
Supplier:   LP ITALIANA
Bottles, water sampling
Catalog # LPIT292258
Supplier:  LP ITALIANA
Bottles, water sampling
Bottles, water sampling

250 ml, Undosed, bulk

Price
617,00 €
Log in for availability
1 * 216 ST
Product Details & Documents

Water sampling bottles, made of PET, graduated, ideal for water sampling and microbiologic analysis.

  • PET bottles with HDPE caps
  • Wide mouth reduces contamination risks
  • Easy to handle due to square section and rounded edges

Cap Ø 50 mm.

Bottles are supplied with or without sodium thiosulphate, when chlorine neutralisation is required.

Specifications
  • Packed:
    bulk
  • Capacity:
    250 ml
  • Seq:
    01
  • Dosage:
    Undosed
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.