Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Mortars and Pestles
Pestles
VWR®, Pestles
VWR®, Pestles
VWR®, Pestles
Supplier:
VWR Collection
Certificates
Expand image
VWR®, Pestles
Supplier:
VWR Collection
About this item
Porcelain.
Grinding surface unglazed
Glazed handle
Certifications:
Produced according to DIN 12906.
Related Information
Documentation
Images 360
410-0122
CoC VWR Pestles
Documentation