Racks for Whirl-Pak® bags
Supplier: NASCO FORT ATKINSON
Certificates
About this item
Storage racks for Whirl-Pak® bags. Perfect for storage and transportation.
- 10 and 12 gauge steel fused together
- Powder coated
- Autoclavable
Rack is finished with an electrostatically applied powder coating. The result is a strong, durable finish which will stand up to many laboratory materials and the rack can be autoclaved. Max dry temperature is 148 °C and wet temperature is 260 °C.