Phusion® High-Fidelity PCR Kit
Supplier: FINNZYMES REAGENTS
Certificates
About this item
Phusion® High-Fidelity PCR Kit offers extreme performance for all major PCR applications. Incorporating Phusion® DNA polymerase, the Phusion® High-Fidelity PCR Kit contains all the reagents necessary, including lambda DNA control template and primers for 1,3 kb and 10 kb amplicons. The template amount is sufficient for performing 20 control reactions in 50 μl volume or 50 control reactions in 20 μl volume.
Delivery: Each kit contains Phusion® DNA polymerase (2 U/µl), 5X Phusion® HF Buffer, 5X Phusion® GC Buffer, dNTP mix (10 mM/each), MgCl₂ solution (50 mM), control lambda template (0,5 ng/µl), 1,3 kb primers (4 µM each), 10 kb primers (4 µM each), and DNA size standard.