Phusion® Flash high-fidelity PCR master mix
Supplier: FINNZYMES REAGENTS
Certificates
About this item
Phusion® Flash High-Fidelity PCR Master Mix is a 2X master mix enabling the use of extremely short PCR protocols without compromising either the fidelity or the yield in the reaction. The master mix utilises Phusion® Flash II DNA Polymerase, a proofreading DNA polymerase modified from Phusion® Hot Start II High-Fidelity DNA Polymerase. The fidelity of the Phusion® Flash II DNA Polymerase is 25-fold greater than Taq.