Phire® Hot Start II DNA polymerase
Supplier: FINNZYMES REAGENTS
Certificates
About this item
Phire® Hot Start II DNA Polymerase is faster, extremely robust, and capable of amplifying long DNA fragments with high yields. Phire® Hot Start II DNA Polymerase incorporates a dsDNA-binding domain which allows short extension times (10 to 15 s/kb), improves yields, and increases fidelity 2-fold compared to Taq DNA polymerase. In addition, the Hot Start technology allows complete reactivation of the enzyme in “zero-time” at standard cycling temperatures. This combination of features makes the polymerase an ideal solution for routine and high throughput PCR applications. Phire® Hot Start II DNA Polymerase delivers superior performance in conventional thermal cyclers as well as in fast instruments, such as the Piko® Thermal Cycler.
- Quick hot start: No reactivation step
- Fast enzyme: Amplify four times faster than with Hot Start Taq
- Robust: Minimal reaction optimisation due to high inhibitor tolerance
- High yields: Abundant products due to high efficiency
- Longer PCR products: Amplify significantly longer DNA fragments than with hot start Taq