Digital desktop timers, Prisma 200 / Prisma 400
Supplier: HANHART
Certificates
About this item
Timers with two (Prisma 200) or four channels (Prisma 400).
- Channels can be started simultaneously
- Addition, Flyback, Countdown
- Adjustable acoustic alarm, clock
- Repeat, manual or automatic
- Selectable: 1/10 second or 1/100 minute
- Timing capacity: 10 hours