Taq DNA polymerase (native, without BSA)
Catalog # EP0282
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 KIT
- Description:Taq DNA Polymerase (native, without BSA), 500 units (5 U/µl)
Specifications
About this item
Taq DNA Polymerase is a highly thermostable DNA polymerase from the thermophilic bacterium Thermus aquaticus. The enzyme catalyses 5'→3' synthesis of DNA, has no detectable 3'→5' exonuclease (proofreading) activity and possesses low 5'→3' exonuclease activity. In addition, Taq DNA polymerase exhibits deoxynucleotidyl transferase activity, which frequently results in the addition of extra adenines at the 3’-end of PCR products. Native Taq DNA polymerase is preferred for amplification of bacterial DNA sequences homologous to those found in E.coli.
- Thermostable with half life more than 40 minutes at 95 °C
- Generates PCR products with 3'-dA overhangs
- Supplied with two buffers, including 10X Taq buffer with (NH₄)₂SO₄, which allows PCR at a wide range of magnesium concentrations and decreases unspecific priming
- Incorporates modified nucleotides (e.g., biotin-, digoxigenin-, fluorescently-labelled nucleotides)
Delivery: Supplied with 10X Taq buffer with KCL, 10X Taq buffer with (NH₄)₂SO₄, and 25 mM MgCl₂.