ContactUsLinkComponent
Centrifuge tubes, conical bottom
Centrifuge tubes, conical bottom
Catalog # WITG4.945.005
Supplier:  witeg Labortechnik
undefined
Centrifuge tubes, conical bottom
Catalog # WITG4.945.005
Supplier:  witeg Labortechnik
Specifications
  • Closure type:
    Without cap
  • Ø×L:
    40×115 mm
  • Capacity:
    85 ml
  • Pk:
    1
Specifications
About this item
DURAN®, borosilicate glass.
  • Heavy wall
  • Ungraduated
  • Very good chemical resistance
  • Very good temperature resistance
Conical bottom, without rim