Liquid samplers, SteriWare®
These disposable Jumbo pipettes are ideal for taking samples from low viscosity liquids. Made from 100% virgin HDPE.
- Quick and easy to use
- Colour-coded tip indicates which end to open and hold
- Cross-contamination risk is eliminated - can be used straight from the bag
- No cleaning is required before or after sampling
- Available as gamma irradiated sterile (SAL 10⁻⁶)
- Suitable for use in ATEX zones 0, 1 and 2 (450 and 900 mm lengths only)
Suitable for all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc).
: FDA acceptable grade conforms to CFR 177.1520.