Orders LinkContactUsLinkComponent
Spatulas, Micro, Disposable, SteriWare®
Spatulas, Micro, Disposable, SteriWare®
Spatulas, Micro, Disposable, SteriWare®
Spatulas, Micro, Disposable, SteriWare®
PS or HDPE, sterile or non sterile. These disposable micro spatulas are ideal for sampling small volumes.
  • Moulded and packed in a cleanroom
  • Suitable for food contact
  • Available sterile (gamma irradiated)
  • Individually packed
The narrow sampling blade enables samples to be taken from very small containers such as vials.
Product Family Options
  • Description
    Sterile
    Length
    Blade length
    Blade width
    Capacity
    Colour
  • PS
    +
    170 mm
    40 mm
    4 mm
    0,3 ml
    White
    1 * 100 ST
    1 * 100 ST - 180,00 €
    180,00 €
    1 * 100 ST
       180,00 €
  • PS
    +
    170 mm
    40 mm
    4 mm
    0,3 ml
    White
    1 * 100 ST
    1 * 100 ST - 402,00 €
    402,00 €
    1 * 100 ST
       402,00 €
  • PS
    -
    170 mm
    40 mm
    6 mm
    0,5 ml
    White
    1 * 100 ST
    1 * 100 ST - 224,00 €
    224,00 €
    1 * 100 ST
       224,00 €
  • PS
    +
    170 mm
    40 mm
    6 mm
    0,5 ml
    White
    1 * 100 ST
    1 * 100 ST - 319,00 €
    319,00 €
    1 * 100 ST
       319,00 €
  • PS
    +
    170 mm
    40 mm
    6 mm
    0,5 ml
    Red
    1 * 100 ST
    1 * 100 ST - 255,00 €
    255,00 €
    1 * 100 ST
       255,00 €
  • PS
    -
    170 mm
    40 mm
    13 mm
    2 ml
    White
    1 * 100 ST
    1 * 100 ST - 164,00 €
    164,00 €
    1 * 100 ST
       164,00 €
  • PS
    +
    170 mm
    40 mm
    13 mm
    2 ml
    White
    1 * 100 ST
    1 * 100 ST - 232,00 €
    232,00 €
    1 * 100 ST
       232,00 €
  • HDPE
    -
    170 mm
    40 mm
    13 mm
    2 ml
    White
    1 * 100 ST
    1 * 100 ST - 240,00 €
    240,00 €
    1 * 100 ST
       240,00 €
  • HDPE
    -
    170 mm
    40 mm
    13 mm
    2 ml
    White
    1 * 100 ST
    1 * 100 ST - 224,00 €
    224,00 €
    1 * 100 ST
       224,00 €