ContactUsLinkComponent
Kjeldahl distillation apparatus
Kjeldahl distillation apparatus
Catalog # WITG2.530.000
Supplier:  witeg Labortechnik
undefined
Kjeldahl distillation apparatus
Catalog # WITG2.530.000
Supplier:  witeg Labortechnik
Specifications
  • Description:
    Apparatus for macro Kjeldahl distillation, complete
  • Flask capacity:
    500 ml
  • Pk:
    1
Specifications
About this item
Kjeldahl distillation unit made of borosilicate glass.
Ordering information: Includes condenser, funnel stopper (ST 10/19), dropping funnel (50 ml), Kjedahl flask (500 ml), adapters and fittings.