Orders LinkContactUsLinkComponent
Sprayer for chromatography
Sprayer for chromatography
Sprayer, with erlenmeyer flask 100 ml, ST 19/26
Catalog # WITG2.147.000
Supplier:   witeg Labortechnik
Sprayer for chromatography
Catalog # WITG2.147.000
Supplier:  witeg Labortechnik
Sprayer for chromatography
Sprayer for chromatography
Sprayer, with erlenmeyer flask 100 ml, ST 19/26
Price
72,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Sprayer are made of borosilicate glass, complete with rubber bulb.
Specifications
  • Description:
    Sprayer, with erlenmeyer flask 100 ml, ST 19/26