Evaporative concentrators, Kuderna-Danish, WHEATON®
WHEA297883
- Max. capacity:500 ml
- Description:Kuderna-Danish concenterator, complete (270 mm synder column, flask, receiving vessel and two clear-seal® joints)
- Pk:1
This unit is designed for the concentration of trace amounts of analytes in an organic solvent prior to analysis.
- Smooth clear-seal joints eliminate the need for grease, reduces sample contamination
- Consists of a 270 mm three-ball Snyder column, a 500 ml flask, and a 10 ml receiving vessel
- Made of WHEATON 33 low extractable borosilicate glass meets USP Type I and ASTM E 438 Type I, Class A requirements