Dropping funnels, cylindrical, without pressure balance
WITG1.856.002
- Cone size:29/32 NS
- Socket size:29/32 NS
- Capacity:2000 ml
- Division:50 ml
- Pk:1
Dropping funnels, made of borosilicate glass 3.3, cylindrical, stopcock with glass key and screw-thread retaling nut.
- Graduated
- With NS cone and socket
- Very good chemical and temperature resistance
: DIN 12567