Condensers, Dimroth, double coils
Catalog # WITG1.750.400
Supplier: witeg Labortechnik
Specifications
- Cone size:24/29 NS
- Socket size:24/29 NS
- Pk:1
Specifications
About this item
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With two cooling coils. Water connection with two glass olives.